Learn More
Motic™ Widefield Eyepiece WF5X/22mm
Widen your field of view for more complete specimen viewing. Motic™ Widefield Eyepiece WF5X/22mm provides an improved field of view and greater eye relief.
Brand: Motic™ 51020141
This item is not returnable.
View return policy
Description
- Widefield style provides improved field of view and greater eye relief.
- Magnification: 5X.
- Field of view: 22mm.
Specifications
Widefield | |
FOV: 22mm | |
ST-39, SFC-11, SMZ-140 Microscopes | |
5X |