Learn More
Ansell™ Winter Hyd-Tuf™ 52-590 Series Brown Nitrile Heavy Weight Gloves
Shop All Ansell Products
Click to view available options
Size:
9
10
Unit Size:
6 pairs
Description
- Nitrile impregnated coating
- Soft, flexible jersey lining
- Absorbs perspiration and offers close, comfortable fit
- Cut and sewn construction
- Cotton jersey liner
- palm impregnated finish
- Safety cuff
Specifications
Specifications
| Back Material | Nitrile |
| Certifications/Compliance | Standard CATEGORY II EN 388 : 3222, EN 511 : 110 |
| Cuff Material | Nitrile |
| Coating Material | Nitrile |
| Product Type | Heavy Weight Gloves |
| Size | 9 |
| Style | 52-590 |
| Glove Cuff Style | Safety |
| Lining | Cotton Jersey |
| Palm Type | Coated-palm |
| Show More |