missing translation for 'onlineSavingsMsg'
Learn More

WIPF1/WIP Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

168.46 EUR - 343.40 EUR

Specifications

Antigen WIPF1/WIP
Dilution Western Blot, Immunohistochemistry 1:100 - 1:200, Immunohistochemistry-Paraffin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18344275
View Documents Promotion Details
Bio-Techne
NBP3-15508-20UL
20 μg
168.46 EUR
20µL
Please to purchase this item. Need a web account? Register with us today!
18331485
View Documents Promotion Details
Bio-Techne
NBP3-15508-100UL
100 μg
343.40 EUR
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

WIPF1/WIP Polyclonal antibody specifically detects WIPF1/WIP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

WIPF1/WIP
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cell Biology, Cytoskeleton Markers, Signal Transduction
PBS, 50% glycerol, pH7.3
7456
IgG
Affinity purified
Western Blot, Immunohistochemistry 1:100 - 1:200, Immunohistochemistry-Paraffin
Polyclonal
Purified
RUO
Human, Mouse, Rat
WAS/WASL interacting protein family, member 1, WAS/WASL-interacting protein family member 1, WASP-interacting protein, WASPIP, WIPMGC111041, WiskProtein PRPL-2, WiskPRPL-2
A synthetic peptide corresponding to a sequence within amino acids 400-500 of human WIPF1/WIP (NP_001070737.1).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications