Learn More
Wooden Dissecting Board With Frame Edges
missing translation for 'orderingAttributeHoverText'
Dimensions (L x W x H):
550 x 400 x 17 mm
missing translation for 'unitSize'
Each
Specifications
Specifications
| Product Type | Dissecting Board |
| Material | Wood |
| Dimensions (L x W x H) | 550 x 400 x 17 mm |
| Height (Metric) | 30 mm |