Learn More
Memmert™ Works Calibration Certificate - Factory 160°C
Product Code. 17120335 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
WTB11
WTB15
WTB24
WTB35
WTB50
WTB6
Unit Size:
Each
Product Code. 17120335
Brand: Memmert™ WTB15D00123
Currently Unavailable: We have no information when this product will be available.
View alternative products
Factory calibration certificate +160°C
  • Accessory
  • Factory calibration certificate
  • 160°C
Specifications
For Use With (Equipment) WTB15
Product Type Works Calibration Certificate