Learn More
Endecotts™ Woven Wire Mesh Sieve
Available in a variety of diameters and apertures
Brand:  Endecotts™ 200SIW.250
 View more versions of this product
Product Code. 11557494
  • 147.00 EUR / Each
Estimated Shipment: 11-03-2025
to see stock.
Explore more special offers
Description
Endecotts Woven Wire Mesh Sieves are made of stainless steel and available in a wide variety of diameters and apertures.
  • Woven Wire Mesh Sieve
  • Made of Stainless Steel
Specifications
Specifications
250 μm
7.87 in.
200 mm
Woven Mesh
Sieve
Stainless Steel
Stainless Steel
4
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products