Learn More
Endecotts™ Woven Wire Mesh Sieve
Product Code. 15631694
Click to view available options
Diameter (Metric) Outer:
200 mm
11.8 in.
300 mm
450 mm
Pore Size:
40 μm
75 μm
1 mm
125 μm
150 μm
2 mm
250 μm
300 μm
425 μm
500 μm
600 μm
Unit Size:
Each
Product Code. 15631694
Brand: Endecotts™ 200SIW.125
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Available in a variety of diameters and apertures
Endecotts Woven Wire Mesh Sieves are made of stainless steel and available in a wide variety of diameters and apertures.
  • Woven Wire Mesh Sieve
  • Made of Stainless Steel
Specifications
Pore Size 125 μm
Type Sieve
Diameter (English) Outer 7.87 in.
Material Stainless Steel
Diameter (Metric) Outer 200 mm
Frame Material Stainless Steel
Mesh Material Woven Mesh
Sieve Number 8