Learn More
Abeba™ X-Light Low Safety Shoe
Product Code. 17583155
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 17583155
Brand: Abeba™ 71103838
Abeba X-Light Low Safety Shoe has a breathable inner lining with silver ions.
  • Size 35 to 48
  • CE, EN ISO 20345:2011, S2, SRC
  • Smooth leather, black
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Slip resistance PU outsole sole
  • Steel toe cap
  • Replaceable Active Comfort Insole (art. 351620)
  • Antistatic
  • Reflective material
Specifications
Closure Type Laces
Color Black
Material Leather
Size 38
Toe Material Steel
Certifications/Compliance (Europe) CE, EN ISO 20345:2011, S2, SRC