Learn More
ABEBA™ X-Light White Lace Up Safety Shoes S2
Product Code. 16328241
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 16328241
Brand: ABEBA™ 7131026/40
ABEBA™ X-Light White Lace Up Safety Shoes S2 with silver ions lining to reduce odor and a replaceable Active Comfort insole.
  • Lightweight, comfort, hygiene
  • Comply to EN ISO 20345 : 2012, S2 SRC
  • ESD dissipative according to EN 61340 Climate class 1
  • Slip resistant PU outsole SRC
  • Good cushioning
  • White washable microfiber
  • Silver ions lining
  • Replaceable Active Comfort insole
  • Sanitized treatment
Specifications
Certifications/Compliance CE EN ISO 20345:2012 S2, SRC, ESD
Closure Type Laces
Color White
Liner Material Breathable lining with Silver ions
Material Microfiber
Size 40
Sole Features Slip and abrasion resistant, shock absorbing, ESD dissipative
Sole Material Polyurethane
Style Lace-up shoes
Toe Material Steel
Packaging Quantity 1 Pair
Certifications/Compliance (Europe) CE EN ISO 20345:2012, DIN EN 61340
Resistant To Water