Learn More
Thermo Scientific™ SS Agar
Click to view available options
Quantity:
10 Plates
Unit Size:
Pack of 10
Description
Enhance the detection of Salmonella and Shigella species with SS Agar, offering superior selectivity and differentiation.
Specifications
Specifications
| Color | Transparent |
| Content And Storage | 2°C to 12°C |
| Format | 90 mm |
| Manufacturing Origin | United Kingdom |
| Product Type | Agar Plate |
| Fill Volume | 17 mL |
| Quantity | 10 Plates |