Learn More
RETSCH XS4002S EasySieve Software Balance
Avoid random errors and operator errors in the evaluation of sieve analysis. Retsch™ XS4002S EasySieve Software Balance is controlled through RS232 serial interface for easy evaluation and documentation of results.
Brand:  RETSCH 32.900.0001
Product Code. 11994412
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
  • For use with Retsch sieve shakers
EasySieve Software
  • Validated according to DIN ISO IEC 12 119
  • Takes over operator guidance and provides instructions about the next working step to be carried out throughout the whole sieving process
  • Reduces the total time required for a sieve analysis by approximately 60 to 70% compared with a manual method
  • Communicates with the balance and sieve shaker and calculates the sieving result as a cumulative distribution and frequency density
  • Permits the calculation of the relevant parameters
  • Graphic and tabular presentations of the sieving result are shown as well as the general sieving data in a well laid out standardized measuring protocol
  • Trend analyses, comparative presentations, mean value calculations and data export are possible
Specifications
Specifications
EasySieve Software Balance XS4002S
200/203 mm
EasySieve Evaluation Software