missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ XY 018
GREENER_CHOICE

Product Code. 18773998
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18773998

Brand: Tocris Bioscience™ 6223/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ROR/NR1F Antagonists

XY 018 is a RORγ antagonist (EC50 = 190 nM). Suppresses androgen receptor expression in prostate cancer cell lines. Also inhibits growth of tumors in androgen receptor-positive xenograft models.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1873358-87-2
Synonym N-[2′-Fluoro-4′-[2,2,2-trifluoro-1-hydroxy-1-(trifluoromethyl)ethyl][1,1′biphenyl]-4-yl]-2-nitrobenzeneacetamide
Quantity 10 mg
Molecular Formula C23H15F7N2O4
Formula Weight 516.36
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.