missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Yeast Extract Glucose Chloramphenicol Agar

Recommended by the International Dairy Federation for enumerating yeasts and molds in milk and milk products. BD Difco Yeast Extract Glucose Chloramphenicol Agar is a selective agar nutrient medium that inhibits the growth of organisms other than yeasts and molds due to the presence of chloramphenicol.

Brand:  BD Difco™ 219001


Product Code. 11718303

  • 108.00 EUR / 500g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The antibiotic method for enumerating yeasts and molds in dairy products has become the method of choice, replacing the traditional acidified method.
  • Results in better recovery of injured fungal cells, which are sensitive to an acid environment
  • Generates less interference from precipitated food particles during the counting
  • Has advantages when a sample contains predominantly yeasts and/or injured yeasts
  • Approximate Formula Per Liter:
    • Yeast Extract: 5.0g
    • Glucose: 20.0g
    • Chloramphenicol: 0.1g
    • Agar: 13.0g
TRUSTED_SUSTAINABILITY
Specifications

Specifications

5 g Yeast Extract, 20g Glucose, 0.1g Chloramphenicol, 13g Agar
Selective Agar Medium
Enumerating yeasts and molds in milk and milk products
500 g
Dehydrated, powder
Chloramphenicol
6.6 ± 0.2
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 1A

Hazard Statement
  • H350-May cause cancer.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with all local/regional/national/internationalregulations.

Supplemental information
  • MIXTURE LIST-Contains : Chloramphenicol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.