missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Yeast Extract Peptone Dextrose Broth
Shop All MP Biomedicals Products
Click to view available options
:
YPD Broth; Weight (Metric): 5 kg; Format: Powder
Description
- YPD (YEPD) is a nutritionally rich medium for the growth of S. cerevisiae, containing dextrose, peptone and yeast extract.
- Dextrose is the energy source.
- Yeast extract and peptone provide carbon, nitrogen, minerals, vitamins, trace ingredients and other essential growth nutrients which stimulate the growth of yeasts.
- Bulk powder.
Specifications
Specifications
| Concentration | 67 g/L |
| Form | Powder |
| Grade | Molecular Biology Certified™ |
| Media Type | YPD |
| Formulation | 20 g Peptone Y; 10 g Yeast Extract-Y; 20 g Dextrose; 17 g Agar-Y. Small quantities of biological buffers have been added for proper pH adjustment. |
| pH | 6.2 ±0.3 (at 25°C) |
| Solubility | Soluble in Water |
| Weight (Metric) | 5 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction