Learn More
ZEOprep™ Diol Functionalized
Shop All Zeochem Products
Click to view available options
:
1 kg
100 g
5 kg
500 g
Description
- DIOL functionalized for low polarity surface
- Ideal for lipid applications
- Suitable for SFC Irregular Amorphous Silica Gel
- High Purity
- European Manufacture
- Swiss accuracy and consistency
- Excellent batch to batch reproducibility
- ISO Certification
- High Chemical and Mechanical Stability
- Narrow Particles Size Distribution
- 20 to 500 μm particles size range
- Available in 60 A or 90 A pore size
- High loadability
- Selection of pack sizes available
- Suitable for gravity, flash, prep and process scale
Specifications
Specifications
| Particle Size | 40 to 63 μm |
| Pore Size | 60 Å |
| Technique | Chromatography |
| Quantity | 500 g |
| Type | Silica Gel |
| Material | Silica Gel |
| Phase | DIOL Functionalized |
| Particle Shape | Irregular |