Learn More
ZFYVE21 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24806-1-AP-150UL
 View more versions of this product
Product Code. 16873305
  • 410.00 EUR / 150µL
Explore more special offers
Description
Description
ZF21 was initially identified as protein that could bind to the cytoplasmic tail of MT1-MMP (Membrane-type 1 matrix metalloproteinase), a potent invasion-promoting protease. ZF21 is a member of a protein family characterized by the presence of a phosphatidylinositol 3-phosphate-binding FYVE domain and regulates focal adhesions (FAs) and cell movement. Knockdown of ZF21 expression resulted in a delay of FA disassembly following induction of synchronous disassembly of FAs by nocodazole treatment, suggesting that ZF21 is involved in FA disassembly. ZF21 contains a noncanonical pleckstrin homology domain that is a possible therapeutic target to treat metastatic cancer.
Specifications
Specifications
ZFYVE21
Polyclonal
Unconjugated
ZFYVE21
ZFYVE21
Rabbit
Antigen Affinity Chromatography
RUO
79038
-20°C
Liquid
Immunoprecipitation, Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BQ24
ZFYVE21
ZFYVE21 Fusion Protein Ag20024
150 μL
Primary
Human
Antibody
IgG