missing translation for 'onlineSavingsMsg'
Learn More

Zinc formate dihydrate, 98%, Alfa Aesar™

Product Code. 11301339
Click to view available options
250g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 5970-62-7
Physical Form Powder
Quantity 250 g
Assay Percent Range 98%
Molecular Formula C2H6O6Zn
MDL Number MFCD00050836
Synonym zinc formate-water 1/2/2, acmc-1at6z, formic acid zinc salt dihydrate, formic acid, zinc salt 2:1, zinc 2+ ion diformate dihydrate, zinc 2+ diformate dihydrate
InChI Key AJWXULAAWUPOJS-UHFFFAOYSA-L
SMILES C(=O)[O-].C(=O)[O-].O.O.[Zn+2]
IUPAC Name zinc;diformate;dihydrate
Molecular Weight (g/mol) 191.444
PubChem CID 57350948
Formula Weight 191.43 (155.40anhydrous)
Density 2.201g/mL
Chemical Name or Material Zinc formate dihydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.