missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Oxide, ReagentPlus™, 99.9%, Honeywell™
Click to view available options
Quantity:
100 g
1 kg
5 kg
Packaging:
Drum with Inliner
Poly Bottle
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Melting Point | 1975°C |
Color | White |
Physical Form | Powder |
Quantity | 100 g |
Linear Formula | ZnO |
UN Number | UN3077 |
Formula Weight | 81.39g/mol |
Percent Purity | 99.90% |
Grade | ReagentPlus™ |
Packaging | Poly Bottle |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'