missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF545 293T Cell Overexpression Lysate (Denatured), Abnova
Brand: Abnova H00284406-T01.100uL
This item is not returnable.
View return policy
Description
Western Blotting
Specifications
293T | |
ZNF545 293T Cell Transient Overexpression Lysate(Denatured) | |
58.63 | |
pCMV-ZNF545 full-length | |
100 μL | |
Human |
Western Blot | |
NM_133466.1 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-ZNF545 antibody by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction