accessibility menu, dialog, popup

Test

CAS RN 1017778-68-5

CH3OFF

CAS RN 1017778-68-5

IUPAC Name: 3,5-difluoro-2-methylbenzaldehyde
Synonyms: 3,5-difluoro-2-methylbenzaldehyde
Molecular Weight (g/mol): 156.13
Molecular Formula: C8H6F2O
InChi Key: MTPAERCJUJOIAH-UHFFFAOYSA-N
SMILES: CC1=C(F)C=C(F)C=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results