accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 103-71-9

CAS RN 103-71-9

IUPAC Name: isocyanatobenzene
Synonyms: phenyl isocyanate phenylcarbimide carbanil PhNCO
Molecular Weight (g/mol): 119.12
Molecular Formula: C7H5NO
InChi Key: DGTNSSLYPYDJGL-UHFFFAOYSA-N
SMILES: O=C=NC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (1)
  • (3)
Boiling Point 
  • (3)
  • (3)

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (3)
  • (3)
  • (5)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.