accessibility menu, dialog, popup

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 103031-30-7

CAS RN 103031-30-7

IUPAC Name: 5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalene-2-carboxylic acid
Synonyms: 5,5,8,8-tetramethyl-6,7-dihydronaphthalene-2-carboxylic acid
Molecular Weight (g/mol): 232.32
Molecular Formula: C15H20O2
InChi Key: KSEZYZSBKCPEKP-UHFFFAOYSA-N
SMILES: CC1(C)CCC(C)(C)C2=C1C=CC(=C2)C(O)=O
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.