accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 103260-44-2

CAS RN 103260-44-2

IUPAC Name: ethyl 2-(oxan-4-yl)acetate
Synonyms: ethyl 2-(oxan-4-yl)acetate
Molecular Weight (g/mol): 172.22
Molecular Formula: C9H16O3
InChi Key: JLMMMEDWRUVCEW-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.