accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1034343-98-0

CAS RN 1034343-98-0

IUPAC Name: methane
Synonyms: methane collocarb carbene, 13c-labeled 13C-labeled carbene kosmos spheron magecol grosafe humenegro furnal
Molecular Weight (g/mol): 16.04
Molecular Formula: CH4
InChi Key: VNWKTOKETHGBQD-UHFFFAOYSA-N
SMILES: C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.