accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10396-10-8

CAS RN 10396-10-8

IUPAC Name: (4-methylbenzenesulfonamido)urea
Synonyms: 4-methylbenzenesulfonamidourea
Molecular Weight (g/mol): 229.25
Molecular Formula: C8H11N3O3S
InChi Key: VRFNYSYURHAPFL-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)NNC(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.