accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10419-77-9

CAS RN 10419-77-9

IUPAC Name: diethyl (iodomethyl)phosphonate
Synonyms: diethyl iodomethylphosphonate
Molecular Weight (g/mol): 278.03
Molecular Formula: C5H12IO3P
InChi Key: PNFHTBDRPOJJTQ-UHFFFAOYSA-N
SMILES: CCOP(=O)(CI)OCC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.