accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 104195-80-4

CAS RN 104195-80-4

IUPAC Name: (2S,3S)-2-azaniumyl-3-methoxybutanoate
Synonyms: (2S,3S)-2-ammonio-3-methoxybutanoate
Molecular Weight (g/mol): 133.15
Molecular Formula: C5H11NO3
InChi Key: FYCWLJLGIAUCCL-IMJSIDKUSA-N
SMILES: CO[C@@H](C)[C@H]([NH3+])C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.