accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 104317-54-6

CAS RN 104317-54-6

IUPAC Name: 3-hydroxybenzene-1-carbothioamide
Synonyms: 3-hydroxybenzenecarbothioamide
Molecular Weight (g/mol): 153.2
Molecular Formula: C7H7NOS
InChi Key: IDIHGRWUQGTNKS-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC(O)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.