accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 105942-09-4

CAS RN 105942-09-4

IUPAC Name: 4-(bromomethyl)-3-fluorobenzonitrile
Synonyms: 4-(bromomethyl)-3-fluorobenzonitrile
Molecular Weight (g/mol): 214.04
Molecular Formula: C8H5BrFN
InChi Key: ZESZAIOGACKOMB-UHFFFAOYSA-N
SMILES: FC1=C(CBr)C=CC(=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.