accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 106-79-6

CAS RN 106-79-6

IUPAC Name: 1,10-dimethyl decanedioate
Synonyms: decanedioic acid, dimethyl ester
Molecular Weight (g/mol): 230.3
Molecular Formula: C12H22O4
InChi Key: ALOUNLDAKADEEB-UHFFFAOYSA-N
SMILES: COC(=O)CCCCCCCCC(=O)OC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.