CAS RN 106719-44-2
CAS RN 106719-44-2
IUPAC Name:
6-amino-2-{[(tert-butoxy)carbonyl]amino}hexanoic acid
Synonyms:
6-amino-2-[(tert-butoxycarbonyl)amino]hexanoic acid
Molecular Weight (g/mol):
246.31
Molecular Formula:
C11H22N2O4
InChi Key:
DQUHYEDEGRNAFO-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)NC(CCCCN)C(O)=O
1
–
1
of
1
results
1
Nalpha-Boc-D-lysine, 98%, Thermo Scientific Chemicals
CAS: 106719-44-2 Molecular Formula: C11H22N2O4 Molecular Weight (g/mol): 246.31 MDL Number: MFCD00076956 InChI Key: DQUHYEDEGRNAFO-SVGMAFHSNA-N Synonym: boc-d-lys-oh,boc-l-lys-oh,n2-boc-d-lysine,nalpha-tert-butoxycarbonyl-d-lysine,nalpha-boc-d-lysine,n-,a-boc-d-lysine,boc-d-lysine,d-lysine, n2-1,1-dimethylethoxy carbonyl,n-tert-butoxycarbonyl-d-lysine,n-alpha-t-butoxycarbonyl-d-lysine PubChem CID: 7018770 IUPAC Name: (2R)-6-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid SMILES: CC(C)(C)OC(=O)N[C@H](CCCCN)C(O)=O