accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 107-07-3

CAS RN 107-07-3

IUPAC Name: 2-chloroethan-1-ol
Synonyms: 2-chloroethanol chloroethylowy alkohol ethylchlorohydrin chlorohydrin ethylene glycol ethylene chlorohydrin (substance) aethylenechlorhydrin ethylene chlorhydrin glycol chlorohydrin 2-cloroetanolo ethene, chlorohydrin
Molecular Weight (g/mol): 80.51
Molecular Formula: C2H5ClO
InChi Key: SZIFAVKTNFCBPC-UHFFFAOYSA-N
SMILES: OCCCl
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (6)
Boiling Point 
  • (4)
  • (3)
Percent Purity 
  • (3)
  • (3)

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)
  • (4)
  • (3)
  • (7)
  • (3)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.