man

Test

CAS RN 36457-58-6

FFFSOOHN

CAS RN 36457-58-6

IUPAC Name: N-benzyl-1,1,1-trifluoromethanesulfonamide
Synonyms: trifluoromethanesulfonamidenbenzyl
Molecular Weight (g/mol): 239.21
Molecular Formula: C8H8F3NO2S
InChi Key: IJHVVEQTOXFGCL-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)NCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results