accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 59025-03-5

CAS RN 59025-03-5

IUPAC Name: 2,5-dioxooxolan-3-yl acetate
Synonyms: 2,5-dioxooxolan-3-yl acetate
Molecular Weight (g/mol): 158.11
Molecular Formula: C6H6O5
InChi Key: SSWJHSASZZAIAU-UHFFFAOYNA-N
SMILES: CC(=O)OC1CC(=O)OC1=O
Quantity 
  • (1)
  • (1)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.