Histology
Products for Your Workflow
Histology is the study of the microscopic anatomy of cells and tissues. Fisher Scientific offers a wide range of chemicals and reagents, including histology-grade solvents, designed specifically for medical laboratory professionals. Let Fisher Scientific help you find everything you need for use in your histology applications. Explore products by technical procedure:
Shop by PCR and qPCR Methods
Sample Preparation
In the fixation stage, a fixative is used to modify tissue by stabilizing the protein, so it is resistant to further changes; thus, fixatives are used to protect tissue from degradation and to maintain the structure of the cells, including sub-cellular components, such as cell organelles (nucleus, endoplasmic reticulum and mitochondria). In the processing stage, tissues must go through a dehydration process to replace water with the material that will be used for embedding, so the tissue samples adhere to the embedding material. The most common technique is paraffin wax embedding.
Samples are immersed in multiple baths of progressively more concentrated ethanol to remove water from the tissues, followed by a clearing agent, such as xylene, to remove the alcohol and finally hot molten paraffin wax, which replaces the xylene and infiltrates the tissue.
Top Categories
Featured Products
Feature product not configured.
Embedding and Sectionning
In the embedding process, tissue samples and liquid embedding material are placed into molds and allowed to harden. The hardened blocks containing the tissue samples are then ready to be sectioned. Embedding can also be accomplished using frozen, non-fixed tissue in a water-based medium, usually a water-based glycol or resin.
In the sectioning phase, the tissue is sliced into very thin (0.5 - 10 microns; 1000 microns = 1mm) sections using a microtome. These slices, usually thinner than the average cell, are then placed on a glass slide for staining. Frozen tissue embedded in a freezing medium is cut on a microtome in a cryostat.
Top Categories
Featured Products
Feature product not configured.
Staining and Imaging
Routine staining is done to give contrast to the tissue being examined. Hematoxylin and Eosin (H&E) are the most commonly used stains in histology.
Hematoxylin is combined with a metal mordant, such as aluminum, and the hematoxylin-aluminum complex is used to stain cell nuclei blue/black. Eosin is the most widely used counterstain in the routine staining of tissue sections. The best staining with eosin occurs at pH 4.5 to 5, and when used properly, at least three shades of pink can be obtained. Erythrocytes, collagen and the cytoplasm of epithelial or muscle cells should stain different shades of pink when using eosin. Samples are immersed in multiple baths of progressively more concentrated ethanol to remove water from the tissues, followed by a clearing agent, such as xylene, to remove the alcohol and finally hot molten paraffin wax, which replaces the xylene and infiltrates the tissue.
Top Categories
Featured Products
Feature product not configured.
Storage
CONTENT
Top Categories
Featured Products
Feature product not configured.
Feature product not configured.
Related Resources
Should You Use Digital or Real-Time PCR?
Should You Use Digital or Real-Time PCR?
Read About PCR Types
Molecular Biology Methods for Disease Research
Molecular Biology Methods for Disease Research
Shop Molecular Biology Products
Custom DNA Oligos Tool
Custom DNA Oligos Tool
Start Shopping Oligonucleotides
Other Applications
Molecular Biology
Molecular Biology Methods
Explore solutions for isolation and purification, amplification, transfection, and more.
Shop Molecular Biology Products
Cell Analysis
Cell Analysis Methods
Find what you need for flow cytometry, cell imaging, and more.
Shop Cell Analysis Products
Cell Culture
Cell Culture
Discover products for seeding, cell expansion, harvesting, and more.
Shop Cell Culture Products
Protein Biology Methods
Protein Biology Methods
Browse isolation, purification, separation, and detection essentials.
Shop Protein Biology Products
Shop by Brand