missing translation for 'onlineSavingsMsg'
Learn More

CD35 Antibody (CR1/8833R), DyLight 488, Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-24014G

545.60 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.


Product Code. 18588856

  • 682.00 EUR / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

CD35 Monoclonal antibody specifically detects CD35 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD35
Monoclonal
DyLight 488
complement component (3b/4b) receptor 1 (Knops blood group), KN
Recombinant fragment (around aa600-900) of human CD35 protein (exact sequence is proprietary)
0.1 mL
Immunology
1378
Store at 4°C in the dark.
IgG κ
Immunohistochemistry (Paraffin)
CR1/8833R
50 mM Sodium Borate
Rabbit
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
CD35 Antibody (CR1/8833R), DyLight 488, Novus Biologicals™ > 0.1 mL, DyLight 488

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.