missing translation for 'onlineSavingsMsg'
Learn More
1,2,4-Benzenetricarboxylic acid, 98%, Thermo Scientific Chemicals Product Code.: 11341877

1,2,4-Benzenetricarboxylic acid, 98%, Thermo Scientific Chemicals

Product Code. 11341877
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11341877

Brand: Thermo Scientific Alfa Aesar L07359.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 528-44-9
Molecular Formula C9H6O6
Molecular Weight (g/mol) 210.14
MDL Number MFCD00002470
InChI Key ARCGXLSVLAOJQL-UHFFFAOYSA-N
Synonym 1,2,4-benzenetricarboxylic acid, trimellitic acid, 1,2,4-tricarboxybenzene, 4-carboxyphthalic acid, 1,3,4-benzenetricarboxylic acid, 1,4,5-benzenetricarboxylic acid, 1,2,4-benzenetricarboxylicacid, unii-7nvy29mq5f, 7nvy29mq5f, dsstox_cid_1487
PubChem CID 10708
ChEBI CHEBI:166055
IUPAC Name benzene-1,2,4-tricarboxylic acid
SMILES OC(=O)C1=CC=C(C(O)=O)C(=C1)C(O)=O

Specifications

Melting Point ∽231°C (decomposition)
Quantity 25 g
Beilstein 2214815
Merck Index 14,9702
Formula Weight 210.14
Percent Purity 98%
Chemical Name or Material 1,2,4-Benzenetricarboxylic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10246249 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberL07359.14
Lot Number10238794 Certificate TypeCertificate of Analysis Date24/02/2023 Catalog NumberL07359.14,L07359.22
Lot NumberQ19H013 Certificate TypeCertificate of Analysis Date13/01/2023 Catalog NumberL07359.14,L07359.22
Lot NumberK13U029 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL07359.14,L07359.22
Lot Number10222202 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL07359.14,L07359.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,2,4-Benzenetricarboxylic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.