missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tri-O-benzoyl-alpha-D-ribofuranose, 97%, Thermo Scientific Chemicals

Product Code. 15435798
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is employed as a starting material for nucleosides and is an intermediate fir cloforabine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a starting material for nucleosides and is an intermediate fir cloforabine.

Solubility
Insoluble in water, easily soluble in chloroform.

Notes
Store at room temperature. Keep away from oxidizing agents and heats.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22224-41-5
Molecular Formula C26H22O8
Molecular Weight (g/mol) 462.454
MDL Number MFCD00080818
InChI Key HUHVPBKTTFVAQF-GCUWJAMBSA-N
Synonym α-D-Ribofuranose 1,3,5-tribenzoate
PubChem CID 68022796
IUPAC Name [(2R,5R)-3,5-dibenzoyloxy-4-hydroxyoxolan-2-yl]methyl benzoate
SMILES C1=CC=C(C=C1)C(=O)OCC2C(C(C(O2)OC(=O)C3=CC=CC=C3)O)OC(=O)C4=CC=CC=C4

Specifications

Melting Point 125°C to 129°C
Color Brown
Quantity 5 g
Beilstein 98446
Solubility Information Insoluble in water,easily soluble in chloroform.
Formula Weight 462.45
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1,3,5-Tri-O-benzoyl-alpha-D-ribofuranose

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,3,5-Tri-O-benzoyl-alpha-D-ribofuranose, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.