Learn More
1,3-Bis(aminopropyl)tetramethyldisiloxane, 94%, Thermo Scientific Chemicals
Description
1,3-Bis(aminopropyl)tetramethyldisiloxane is used as a curing agent for epoxy molding compounds, which is utilized for high-reliability semiconductor devices. It is also used to prepare siloxane- urea copymers from 4,4’-diphenylmethane diisocyanate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,3-Bis(aminopropyl)tetramethyldisiloxane is used as a curing agent for epoxy molding compounds, which is utilized for high-reliability semiconductor devices. It is also used to prepare siloxane- urea copymers from 4,4’-diphenylmethane diisocyanate.
Solubility
Slightly soluble in water.
Notes
Air Sensitive. Incompatible with strong oxidizing agents. Store in a cool place.

Specifications
Specifications
Boiling Point | 132°C to 139°C (11mmHg) |
Quantity | 25 g |
UN Number | UN2735 |
Beilstein | 1760876 |
Flash Point | 91°C (195°F) |
Solubility Information | Slightly soluble in water. |
Formula Weight | 248.52 |
Percent Purity | 94% |
Refractive Index | 1.45 |
Sensitivity | Air Sensitive |
Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.