Learn More
1,4-Phenylenediacetonitrile, 97%, Thermo Scientific Chemicals
Description
1,4-Phenylenediacetonitrile, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,4-Phenylenediacetonitrile, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Specifications
Specifications
Melting Point | 94°C to 98°C |
Quantity | 5 g |
UN Number | UN3439 |
Beilstein | 1866157 |
Solubility Information | Insoluble in water. |
Formula Weight | 156.19 |
Percent Purity | 97% |
Chemical Name or Material | 1,4-Phenylenediacetonitrile |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10235634 | Certificate of Analysis | 07/12/2021 | L13464.06,L13464.14 |
10180259 | Certificate of Analysis | 06/12/2021 | L13464.06,L13464.14 |
10144961 | Certificate of Analysis | 06/12/2021 | L13464.06,L13464.14 |
10225141 | Certificate of Analysis | 06/12/2021 | L13464.06,L13464.14 |
10202420 | Certificate of Analysis | 06/12/2021 | L13464.06,L13464.14 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.