Learn More
1,6-Bismaleimidohexane, 97%, Thermo Scientific Chemicals
156.78 EUR - 612.05 EUR
Chemical Identifiers
CAS | 4856-87-5 |
---|---|
Molecular Formula | C14H16N2O4 |
Molecular Weight (g/mol) | 276.292 |
MDL Number | MFCD00047122 |
InChI Key | PYVHLZLQVWXBDZ-UHFFFAOYSA-N |
Synonym | 1,6-bismaleimidohexane, 1,6-dimaleimidohexane, n,n'-hexamethylenedimaleimide, 1,6-bis maleimido hexane, 1,1'-hexane-1,6-diyl bis 1h-pyrrole-2,5-dione, 1h-pyrrole-2,5-dione, 1,1'-1,6-hexanediyl bis, maleimide, n,n'-hexamethylenedi, n,n'-hexamethylenebis maleimide, 1,1'-1,6-hexanediyl bis-1h-pyrrole-2,5-dioneShow More |
PubChem CID | 20992 |
IUPAC Name | 1-[6-(2,5-dioxopyrrol-1-yl)hexyl]pyrrole-2,5-dione |
SMILES | C1=CC(=O)N(C1=O)CCCCCCN2C(=O)C=CC2=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15465058
|
Thermo Scientific Alfa Aesar
H58874.MC |
100 mg |
156.78 EUR
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15475058
|
Thermo Scientific Alfa Aesar
H58874.ME |
500 mg |
612.05 EUR
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A sulfhydryl reactive homobifunctional cross linking reagent. Under mild conditions, will permit irreversible cross-linking of sulfhydral containing compounds at a pH range of 6.5 to 7.5.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA sulfhydryl reactive homobifunctional cross linking reagent. Under mild conditions, will permit irreversible cross-linking of sulfhydral containing compounds at a pH range of 6.5 to 7.5.
Solubility
Soluble in chloroform.
Notes
Protect from heat. Store away from oxidizing agents.
Chemical Identifiers
4856-87-5 | |
276.292 | |
PYVHLZLQVWXBDZ-UHFFFAOYSA-N | |
20992 | |
C1=CC(=O)N(C1=O)CCCCCCN2C(=O)C=CC2=O |
C14H16N2O4 | |
MFCD00047122 | |
1,6-bismaleimidohexane, 1,6-dimaleimidohexane, n,n'-hexamethylenedimaleimide, 1,6-bis maleimido hexane, 1,1'-hexane-1,6-diyl bis 1h-pyrrole-2,5-dione, 1h-pyrrole-2,5-dione, 1,1'-1,6-hexanediyl bis, maleimide, n,n'-hexamethylenedi, n,n'-hexamethylenebis maleimide, 1,1'-1,6-hexanediyl bis-1h-pyrrole-2,5-dioneShow More | |
1-[6-(2,5-dioxopyrrol-1-yl)hexyl]pyrrole-2,5-dione |
Specifications
4856-87-5 | |
C14H16N2O4 | |
1,6-bismaleimidohexane, 1,6-dimaleimidohexane, n,n'-hexamethylenedimaleimide, 1,6-bis maleimido hexane, 1,1'-hexane-1,6-diyl bis 1h-pyrrole-2,5-dione, 1h-pyrrole-2,5-dione, 1,1'-1,6-hexanediyl bis, maleimide, n,n'-hexamethylenedi, n,n'-hexamethylenebis maleimide, 1,1'-1,6-hexanediyl bis-1h-pyrrole-2,5-dione | |
PYVHLZLQVWXBDZ-UHFFFAOYSA-N | |
1-[6-(2,5-dioxopyrrol-1-yl)hexyl]pyrrole-2,5-dione | |
20992 | |
97% |
144°C | |
MFCD00047122 | |
Soluble in chloroform. | |
C1=CC(=O)N(C1=O)CCCCCCN2C(=O)C=CC2=O | |
276.292 | |
276.29 | |
1,6-Bismaleimidohexane |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 225-449-1
missing translation for 'rtecsNumber' : ON5520000
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.