missing translation for 'onlineSavingsMsg'
Learn More
1-Amino-3-(4-methoxyphenoxy)propan-2-ol, ≥95%, Thermo Scientific™ Product Code.: 10582272

1-Amino-3-(4-methoxyphenoxy)propan-2-ol, ≥95%, Thermo Scientific™

Product Code. 10582272
10 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10582272

Brand: Thermo Scientific Maybridge BTB12133EA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5002-93-7
Molecular Formula C10H15NO3
Molecular Weight (g/mol) 197.234
MDL Number MFCD00205693
InChI Key KFQPRNVTVMCYEH-UHFFFAOYSA-N
Synonym 1-amino-3-4-methoxyphenoxy propan-2-ol, 1-amino-3-4-methoxy-phenoxy-propan-2-ol, 2-propanol, 1-amino-3-4-methoxyphenoxy, 3-amino-1-4-methoxyphenoxy propan-2-ol, maybridge1_002199, 4-3-amino-2-hydroxypropoxy anisole, 2rs-1-amino-3-4methoxyphenoxy-2-propanol, 2rs-1-amino-3-4-methoxyphenoxy-2-propanol
PubChem CID 2735360
IUPAC Name 1-amino-3-(4-methoxyphenoxy)propan-2-ol
SMILES COC1=CC=C(C=C1)OCC(CN)O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95+%
Packaging Amber Glass Bottle
Linear Formula CH3OC6H4OCH2CH(OH)CH2NH2
Quantity 10 g
Formula Weight 197.23
Percent Purity 95%
Chemical Name or Material 1-Amino-3-(4-methoxyphenoxy)propan-2-ol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Amino-3-(4-methoxyphenoxy)propan-2-ol, ≥95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.