missing translation for 'onlineSavingsMsg'
Learn More
1-Bromo-3-chloropropane, 99%, Thermo Scientific Chemicals Product Code.: 10646754

1-Bromo-3-chloropropane, 99%, Thermo Scientific Chemicals

Product Code. 10646754
2.5 L, Glass bottle
Click to view available options
Quantity:
50 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10646754

Brand: Thermo Scientific Acros 106860025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-70-6
Molecular Formula C3H6BrCl
Molecular Weight (g/mol) 157.44
MDL Number MFCD00000998
InChI Key MFESCIUQSIBMSM-UHFFFAOYSA-N
Synonym trimethylene chlorobromide, 1-chloro-3-bromopropane, 3-bromopropyl chloride, 3-chloropropyl bromide, propane, 1-bromo-3-chloro, 3-chloro-1-bromopropane, 3-bromo-1-chloropropane, trimethylene bromide chloride, 1,3-chbp, 1,3-chlorbromopropane
PubChem CID 8006
IUPAC Name 1-bromo-3-chloropropane
SMILES C(CCl)CBr

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -59.0°C
Density 1.5900g/mL
Boiling Point 142.0°C to 145.0°C
Flash Point 81°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula Cl(CH2)3Br
Refractive Index 1.4850 to 1.4870
Quantity 2.5 L
Beilstein 01, 109
Specific Gravity 1.59
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol,ethanol,ether and chloroform
Formula Weight 157.44
Percent Purity 99%
Chemical Name or Material 1-Bromo-3-chloropropane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Bromo-3-chloropropane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Germ cell mutagenicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Bromo-3-chloropropane, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.