missing translation for 'onlineSavingsMsg'
Learn More
1-Bromo-4-n-hexylbenzene, 97%, Thermo Scientific Chemicals Product Code.: 11408456

1-Bromo-4-n-hexylbenzene, 97%, Thermo Scientific Chemicals

Product Code. 11408456
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11408456

Brand: Thermo Scientific Alfa Aesar A15918.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23703-22-2
Molecular Formula C12H17Br
Molecular Weight (g/mol) 241.17
MDL Number MFCD00061114
InChI Key MXHOLIARBWJKCR-UHFFFAOYSA-N
Synonym 1-4-bromophenyl hexane, 4-hexylbromobenzene, 1-bromo-4-n-hexylbenzene, 4-bromo-1-hexylbenzene, benzene, 1-bromo-4-hexyl, 4-bromo-n-hexylbenzene, 4-hexylphenyl bromide, 1-bromo-4-hexyl-benzene, acmc-1cb1g, #Show MoreShow Less
PubChem CID 598186
IUPAC Name 1-bromo-4-hexylbenzene
SMILES CCCCCCC1=CC=C(Br)C=C1

Specifications

Density 1.179
Boiling Point 280°C
Flash Point >120°C (248°F)
Refractive Index 1.5213
Quantity 1 g
Formula Weight 241.17
Percent Purity 97%
Chemical Name or Material 1-Bromo-4-n-hexylbenzene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10204815 Certificate TypeCertificate of Analysis Date07/08/2023 Catalog NumberA15918.03,A15918.14,A15918.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Bromo-4-n-hexylbenzene, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.