missing translation for 'onlineSavingsMsg'
Learn More
1-Butanol, ExpertQ, Scharlab Product Code.: 11329923
compliance-icons compliance-icons compliance-icons

1-Butanol, ExpertQ, Scharlab

Product Code. 11329923
1 L
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy

Product Code. 11329923

Brand: Scharlab AL01731000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 71-36-3
Quantity 1 L
Molecular Formula C4H10O
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
SMILES CCCCO
IUPAC Name butan-1-ol
Molecular Weight (g/mol) 74.12
Grade ExpertQ
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Butanol, reagent grade, ACS, ISO, Reag. Ph Eur
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Butanol, ExpertQ, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.