missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloro-3-(trifluoromethoxy)benzene, 98%, Thermo Scientific™
Click to view available options
Quantity:
25 g
100 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
1-Chloro-3-(trifluoromethoxy)benzene is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
1-Chloro-3-(trifluoromethoxy)benzene is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.

Specifications
Specifications
Boiling Point | 140°C |
Quantity | 25 g |
Refractive Index | 1.436 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 196.55 |
Percent Purity | 98% |
Chemical Name or Material | 1-Chloro-3-(trifluoromethoxy)benzene |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
1-Chloro-3-(trifluoromethoxy)benzene, 98%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction