missing translation for 'onlineSavingsMsg'
Learn More
1-Chloronaphthalene, tech. 85%, remainder 2-chloronaphthalene, Thermo Scientific Chemicals Product Code.: 11417277

1-Chloronaphthalene, tech. 85%, remainder 2-chloronaphthalene, Thermo Scientific Chemicals

Product Code. 11417277
1000 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11417277

Brand: Thermo Scientific Alfa Aesar A15029.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-Chloronaphthalene was used as solvent additive in the fabrication of fullerene based photovoltaic devices. It was used in preparation of polyvinyl chloride based membranes of 5,10,15,20-tetrakis(4-methoxyphenyl)porphyrinatocobalt(II) for arsenite determination. Also used in the synthesis of pharmaceutical compounds such as aryl chlorides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloronaphthalene was used as solvent additive in the fabrication of fullerene based photovoltaic devices. It was used in preparation of polyvinyl chloride based membranes of 5,10,15,20-tetrakis(4-methoxyphenyl)porphyrinatocobalt(II) for arsenite determination. Also used in the synthesis of pharmaceutical compounds such as aryl chlorides.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-13-1
Molecular Formula C10H7Cl
Molecular Weight (g/mol) 162.616
MDL Number MFCD00003874
InChI Key JTPNRXUCIXHOKM-UHFFFAOYSA-N
Synonym alpha-chloronaphthalene, naphthalene, 1-chloro, 1-naphthyl chloride, chloronaphthalene, 1-chloro-naphthalene, alpha-naphthyl chloride, 1-chlornaftalen, 1-chloro naphthalene, alpha-chlornaphthalene, .alpha.-chloronaphthalene
PubChem CID 7003
IUPAC Name 1-chloronaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2Cl

Specifications

Melting Point ∼-8°C
Density 1.194
Boiling Point 259°C
Flash Point 121°C (249°F)
Refractive Index 1.633
Quantity 1000 g
Beilstein 970836
Merck Index 14,2149
Solubility Information Insoluble in water.
Formula Weight 162.62
Percent Purity 85%
Grade Technical
Assay remainder 2-chloronaphthalene
Chemical Name or Material 1-Chloronaphthalene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253286 Certificate TypeCertificate of Analysis Date03/12/2024 Catalog NumberA15029.18,A15029.0B,A15029.30
Lot Number10212816 Certificate TypeCertificate of Analysis Date06/06/2023 Catalog NumberA15029.0B
Lot NumberM23J049 Certificate TypeCertificate of Analysis Date27/01/2023 Catalog NumberA15029.18,A15029.0B,A15029.30
Lot NumberM13J024 Certificate TypeCertificate of Analysis Date19/01/2023 Catalog NumberA15029.18,A15029.0B,A15029.30
Lot NumberM13J020 Certificate TypeCertificate of Analysis Date19/01/2023 Catalog NumberA15029.18,A15029.0B,A15029.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Chloronaphthalene, tech. 85%, remainder 2-chloronaphthalene, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.