missing translation for 'onlineSavingsMsg'
Learn More

Fisher Chemical 1 LT DICHLOROMETHANE 1L

Brand:  Fisher Chemical D/1852/PB15X


1 LT DICHLOROMETHANE 1L

Product Code. 11433373

  • 24.45 EUR / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Dichloromethane

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 3

SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2531041 Certificate TypeCertificate of Analysis Date17/03/2025 Catalog NumberD/1852/PB15X,D/1852/17,D/1852/27,D/1852/17Z,D/1852/25,D/1852/15Z,D/1852/PC15
Lot Number2529168 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberD/1852/PB15X,D/1852/17,D/1852/27,D/1852/17Z,D/1852/25,D/1852/15Z,D/1852/PC15
Lot Number2529052 Certificate TypeCertificate of Analysis Date28/02/2025 Catalog NumberD/1852/PB15X,D/1852/17,D/1852/27,D/1852/17Z,D/1852/25,D/1852/15Z,D/1852/PC15
Lot Number2526194 Certificate TypeCertificate of Analysis Date07/02/2025 Catalog NumberD/1852/PB15X,D/1852/17,D/1852/17X,D/1852/27,D/1852/15,D/1852/17Z,D/1852/25,D/1852/24,D/1852/15Z,D/1852/PB17,D/1852/PB15,D/1852/21,D/1852/PC15
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisher Chemical 1 LT DICHLOROMETHANE 1L >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.