missing translation for 'onlineSavingsMsg'
Learn More
1-Methyl-2-pyrrolidinone, Biograde, 99.5%, Thermo Scientific Chemicals Product Code.: 11345138

1-Methyl-2-pyrrolidinone, Biograde, 99.5%, Thermo Scientific Chemicals

Product Code. 11345138
4 L, Under Argon
Click to view available options
Quantity:
1 L
4 L
4 x 4 L
Packaging:
Under Argon
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11345138

Brand: Thermo Scientific Alfa Aesar 044063.K7

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Suitable for Biotech applications.Useful for spectrophotometry, chromatography and ICP-MS detection. Also, used for the chemical functionalization of graphene sheets by solvothermal reduction of graphene oxide.

Suitable for Biotech applications.1-methyl-2-pyrrolidinone, is useful for spectrophotometry, chromatography and ICP-MS detection. It is useful for the chemical functionalization of graphene sheets by solvothermal reduction of graphene oxide. It is also useful for pharmaceutical and different industrial applications due to its broad solvency and low volatility.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications

  • Suitable for Biotech applications
  • Useful for spectrophotometry, chromatography and ICP-MS detection
  • Used for the chemical functionalization of graphene sheets by solvothermal reduction of graphene oxide
  • Also useful for pharmaceutical, different industrial applications due to its broad solvency and low volatility
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-50-4
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
MDL Number MFCD00003193
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
IUPAC Name 1-methylpyrrolidin-2-one
SMILES CN1CCCC1=O

Specifications

Melting Point -24°C
Color Colorless
Density 1.033 g/cm3 at 20°C
Boiling Point 204°C
Quantity 4 L
Flash Point 86°C (187°F)
Ignition Point 270°C (518°F)
Odor Amine-like
Packaging Under Argon
Physical Form Liquid,packaged under Argon
Refractive Index 1.4705
Beilstein 106420
Solubility Information Miscible with water,lower alcohols,ketones,ethyl acetate,castor oil,chloroform and benzene. Moderately miscible with aliphatic hydrocarbons.
Formula Weight 99.13
Grade Biochemical
Chemical Name or Material 1-Methyl-2-pyrrolidinone
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberJ05U909 Certificate TypeCertificate of Analysis Date12/01/2023 Catalog Number044063.K7,044063.M6,044063.K2
Lot NumberQ21H726 Certificate TypeCertificate of Analysis Date21/05/2022 Catalog Number044063.K7,044063.M6,044063.K2
Lot NumberX30H855 Certificate TypeCertificate of Analysis Date27/04/2022 Catalog Number044063.K7,044063.M6,044063.K2
Lot NumberX30H842 Certificate TypeCertificate of Analysis Date27/04/2022 Catalog Number044063.K7,044063.M6,044063.K2
Lot NumberB22R010 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number044063.K7,044063.M6,044063.K2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Methyl-2-pyrrolidinone, Biograde, 99.5%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.