Learn More
Fisherbrand™ 13mm PP Screw Cap, 13-425 thread
Polypropylene caps are chemically inert and suitable for most chromatography applications.
Brand: Fisherbrand™ 13 08 0336
84.74 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
- 13mm
- PP Screw Cap
- Closed Top Screw Cap
Specifications
Black | |
Polypropylene | |
ND13 |
13 mm | |
Closed Top Screw Cap |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 12
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
45Hours
22Minutes
43Seconds
36New Offer Save Up to 70%
Product Content Correction
Product Title
Fisherbrand™ 13mm PP Screw Cap, 13-425 thread > Closed top, Black
Spot an opportunity for improvement?Share a Content Correction